Alnico Bar Magnets
Catalog # 470349-636
Supplier: United Scientific Supplies
Specifications
- Description:Alnico bar magnet pair
- Environmentally Preferable:
- Cat. No.:470349-636
Specifications
About this item
Strong Alnico bar magnets for demonstrations.
- Includes a keeper
- Strong Alnico magnets
- Perfect for demonstrating magnetic fields
Powerful Alnico 1 bar magnets, shipped in pairs, with end keepers. Magnets are painted with North and South poles, clearly marked. Each bar is 2" long and ¹/₄" thick.