Blue-Violet Laser Pointer
Blue-violet laser pointer
Catalog # 470343-198
Supplier: American Scientific
Supplier #: 5134-10
Jump to:
Product Details & Documents
Excite your students with this affordable blue-violet laser pointer.
- Maximum power: <5 mW (class 3R/IIIa)
- Approximate wavelength: 405 nm
- Comes with instructions and 2 AA batteries in a reusable gift box
Violet laser pointers are a bit unusual, so they have that wow-factor for the students. While this is still a class IIIa laser, it appears to be slightly less bright than a green laser pointer. This laser is perfect for wavelength comparisons in diffraction experiments.
Specifications
- Description:Blue-violet laser pointer



