Order Entry
United States
Orders LinkContactUsLinkComponent
Red Laser Pointer

Red Laser Pointer

Red laser pointer
Catalog # 470343-200
Supplier:   American Scientific
Supplier #:  5135-00
Red Laser Pointer
Catalog # 470343-200
Red Laser Pointer
Red Laser Pointer

Red laser pointer

Price
$20.35
Log in for availability
Each
Product Details & Documents

Affordable pen-size red laser pointer.

  • Maximum power: <5 mW (class 3R/IIIa)
  • Approximate wavelength: 630 to 650 nm
  • Includes a teacher’s guide
  • Includes 2 AA batteries in a reusable gift box

This laser pointer is perfect for any number of classroom experiments, such as lenses, prisms, mirrors, and diffraction gratings.

Specifications
  • Description:
    Red laser pointer
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.