Orders LinkContactUsLinkComponent
Vector PCX Derivatization Instruments
Marketsource
Supplier:   Pickering Labs
Vector PCX Derivatization Instruments
Supplier:  Pickering Labs
Marketsource
Certificates
Vector PCX Derivatization Instruments
12 Options Available Below
Product Details & Documents
Vector PCX provides the selectivity and sensitivity required for most standard post-column applications while being reliable and easy to use. Since Vector PCX does not have a column oven it is important to use the HPLC column oven to ensure stable column temperature and prevent retention time drifts and separation problems.
  • Independently adjustable, low-pulsation inert pumps
  • PEEK liquid manifolds, Integrated gas manifold
  • Easy access and monitoring of fluidics
  • Simple control interface (no computer required)
  • Reduced bench space profile
  • Sapphire pistons with automatic piston wash
Dimensions: 43×21.6×41.2 cm (17×8.75×16 inches). Weight: 12.5 kg (27.6 lbs) – Duplex. Electrical: 100 to 120 V, 50/60 Hz 1.7 A, 200 W or 200 to 240 V, 50/60 Hz, 0.8 A, 200 W. Mains voltage ±10% of nominal. Installation (over voltage) category II, pollution degree 2. Environmental: altitude up to 6500 ft. Ambient temperature 15 to 25 ºC. Relative humidity up to 80% at 31 ºC. Adjustable from 0.05 to 2.00 ml/minute against back-pressures of up to 2000 psi.
One liter capacity (2 L reservoirs available). Maintained under inert gas pressure to inhibit oxidation of oxygensensitive reagents. Valve built into reservoir cap permits sparging during reagent preparation. Reagent reservoirs fitted with 3.1 mm (¹/₈") Ext.Ø, oxygen-impermeable Air Barrier tubing for oxygen-sensitive reagents and/or with 3.1 mm (¹/₈") Ext.Ø FEP tubing.
Product Family Options
Single Pump
Product Information
  • Availability
    Price
  • MSPP-1154-4011
    Vector, single pump, OPA flow restrictor, 0.15 ml, 120 V (primary amino acids w/OPA, aminoglycoside antibiotics, biogenic amines, sulfonamide artificial sweetener)
    Vector, single pump, OPA flow restrictor, 0.15 ml, 120 V (primary amino acids w/OPA, aminoglycoside antibiotics, biogenic amines, sulfonamide artificial sweetener)
    Log in for availability
    Log in for availability
    Each
    Each - $29,661.54
    $29,661.54
    Each
       $29,661.54
    Supplier Number: 1154-4011
    Specifications:
    Marketsource
    Marketsource
  • MSPP-1154-4021
    Vector, single pump Trione flow restrictor, 0.5 ml, 120 V (amino acids w/TRIONE®, Theanine)
    Vector, single pump Trione flow restrictor, 0.5 ml, 120 V (amino acids w/TRIONE®, Theanine)
    Log in for availability
    Log in for availability
    Each
    Each - $29,661.54
    $29,661.54
    Each
       $29,661.54
    Supplier Number: 1154-4021
    Specifications:
    Marketsource
    Marketsource
  • MSPP-1154-4103
    Vector, single pump, OPA flow restrictor, 0.5 ml, 120 V (formaldehyde, B vitamins)
    Vector, single pump, OPA flow restrictor, 0.5 ml, 120 V (formaldehyde, B vitamins)
    Log in for availability
    Log in for availability
    Each
    Each - $29,661.54
    $29,661.54
    Each
       $29,661.54
    Supplier Number: 1154-4103
    Specifications:
    Marketsource
    Marketsource
  • MSPP-1154-4031
    Vector, single pump, opa flow restrictor, knitted 1.4 ml, 120 V (aflatoxins, sulfonamide drugs)
    Vector, single pump, opa flow restrictor, knitted 1.4 ml, 120 V (aflatoxins, sulfonamide drugs)
    Log in for availability
    Log in for availability
    Each
    Each - $29,661.54
    $29,661.54
    Each
       $29,661.54
    Supplier Number: 1154-4031
    Specifications:
    Marketsource
    Marketsource
  • MSPP-1154-4101
    Vector, single pump, OPA flow restrictor, knitted 3.5 ml, 120 V (Voglibose)
    Vector, single pump, OPA flow restrictor, knitted 3.5 ml, 120 V (Voglibose)
    Log in for availability
    Log in for availability
    Each
    Each - $29,661.54
    $29,661.54
    Each
       $29,661.54
    Supplier Number: 1154-4101
    Specifications:
    Marketsource
    Marketsource
  • MSPP-1154-4091
    Vector, single pump, custom configuration, 120 V
    Vector, single pump, custom configuration, 120 V
    Log in for availability
    Log in for availability
    Each
    Each - $29,661.54
    $29,661.54
    Each
       $29,661.54
    Supplier Number: 1154-4091
    Specifications:
    Marketsource
    Marketsource
Dual Pump
Product Information
  • Availability
    Price
  • MSPP-1154-4001
    Vector, dual pump, 0.5 ml, 120 V (streptomycin)
    Vector, dual pump, 0.5 ml, 120 V (streptomycin)
    Log in for availability
    Log in for availability
    Each
    Each - $34,830.77
    $34,830.77
    Each
       $34,830.77
    Supplier Number: 1154-4001
    Specifications:
    Marketsource
    Marketsource
  • MSPP-1154-4051
    Vector, dual pump, OPA flow restrictors, 0.5 ml, 120 V (carbamate, glyphosate, primary and secondary amino acids w/OPA, streptomycin)
    Vector, dual pump, OPA flow restrictors, 0.5 ml, 120 V (carbamate, glyphosate, primary and secondary amino acids w/OPA, streptomycin)
    Log in for availability
    Log in for availability
    Each
    Each - $34,830.77
    $34,830.77
    Each
       $34,830.77
    Supplier Number: 1154-4051
    Specifications:
    Marketsource
    Marketsource
  • MSPP-1154-4061
    Vector, dual pump, opa flow restrictors, knitted 1.0 ml, 120 V (paralytic shellfish toxins)
    Vector, dual pump, opa flow restrictors, knitted 1.0 ml, 120 V (paralytic shellfish toxins)
    Log in for availability
    Log in for availability
    Each
    Each - $34,830.77
    $34,830.77
    Each
       $34,830.77
    Supplier Number: 1154-4061
    Specifications:
    Marketsource
    Marketsource
  • MSPP-1154-4071
    Vector, dual pump, OPA flow restrictors, knitted 1.2 and 1.6 ml, 120 V (Tricothescene)
    Vector, dual pump, OPA flow restrictors, knitted 1.2 and 1.6 ml, 120 V (Tricothescene)
    Log in for availability
    Log in for availability
    Each
    Each - $35,323.08
    $35,323.08
    Each
       $35,323.08
    Supplier Number: 1154-4071
    Specifications:
    Marketsource
    Marketsource
  • MSPP-1154-4081
    Vector, dual pump, reverse plumbing, PEEK® tubing flow restrictors, knitted 1.4 ml, 120 V (polyether antibiotics)
    Vector, dual pump, reverse plumbing, PEEK® tubing flow restrictors, knitted 1.4 ml, 120 V (polyether antibiotics)
    Log in for availability
    Log in for availability
    Each
    Each - $34,830.77
    $34,830.77
    Each
       $34,830.77
    Supplier Number: 1154-4081
    Specifications:
    Marketsource
    Marketsource
  • MSPP-1154-4092
    Vector, dual pump, custom configuration, 120 V
    Vector, dual pump, custom configuration, 120 V
    Log in for availability
    Log in for availability
    Each
    Each - $34,830.77
    $34,830.77
    Each
       $34,830.77
    Supplier Number: 1154-4092
    Specifications:
    Marketsource
    Marketsource