ContactUsLinkComponent
Advanced Astronomy Kit
Advanced Astronomy Kit
Catalog # 470350-122
Supplier:  Avantor
undefined
Advanced Astronomy Kit
Catalog # 470350-122
Supplier:  Avantor
Specifications
  • Description:
    Advanced astronomy kit
  • Environmentally Preferable:
  • Cat. No.:
    470350-122
Specifications
About this item
Includes a celestial sphere, a refracting telescope, and a spectrometer with activities.
  • Designed to supplement laboratory work or lecture, or as a home lab
  • Includes an 18 page astronomical instrument kit, manual, and assembly instructions
  • Questions for homework or group discussion paired with each device