Sodium hydroxide, micropearls, extra pure
:1310-73-2
:
:NaOH
:MFCD00003548
:40 g/mol
:1390 °C (760 mmHg)
:318 °C
:Ambient
:1823
:8,II
:GHS05
- Appearance:White Pearls
- Particle size:20 to 40 mesh
- Titration with HCl:≥97.5 % (NaOH)
Frequently Bought Together