


Carbon disulfide 99.9% ACS
Catalog # TS44703-5000
Supplier: Thermo Scientific Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:500 ml
- Cat. No.:TS44703-5000
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Water:≤0.05 %
- GC:≥99.9 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.002 %
- Sulfur dioxide (SO2):≤2.5 ppm
- Hydrogen sulfide (H2S):≤1.5 ppm
Specification Test Results
About this item
:
:76.14 g/mol
Boiling Pt:46 °C (760 mmHg)
Melting Pt:–111 °C
Density:1.262 g/cm³
Flash Pt:–30 °C
UN:1131
ADR:3,I
:GHS07
:GHS08
:GHS02
Storage Temperature:Ambient
CAS Number:75-15-0
Formula:CS₂
MDL Number:MFCD00011321