

1-Chlorobutane 99.5% for analysis
Catalog # TS44702-0010
Supplier: Thermo Scientific Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:1 L
- Cat. No.:TS44702-0010
Specifications
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4010 to 1.4030 (20 °C, 589 nm)
- Water:≤0.05 %
- GC:≥99.4 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.001 %
- Acidity:≤0.0005 meq/g
- Cadmium (Cd):≤0.05 ppm
- Calcium (Ca):≤0.5 ppm
- Chromium (Cr):≤0.02 ppm
- Cobalt (Co):≤0.02 ppm
- Copper (Cu):≤0.02 ppm
- Iron (Fe):≤0.1 ppm
- Lead (Pb):≤0.1 ppm
- Magnesium (Mg):≤0.1 ppm
- Manganese (Mn):≤0.02 ppm
- Nickel (Ni):≤0.02 ppm
- Zinc (Zn):≤0.1 ppm
- Sulfur compounds (as S):≤0.002 %
Specification Test Results
About this item
:
:92.57 g/mol
Boiling Pt:77…78 °C (760 mmHg)
Melting Pt:–123 °C
Density:0.88 g/cm³
Flash Pt:–12 °C
UN:1127
ADR:3,II
:GHS02
:GHS08
Storage Temperature:Ambient
CAS Number:109-69-3
Formula:CH₃(CH₂)₃Cl
MDL Number:MFCD00001009