ContactUsLinkComponent
ProPlus PhyTip® Columns, Biotage
ProPlus PhyTip® Columns, Biotage
Catalog # MSPP-PTB922007
Supplier:  BIOTAGE LLC MS
ProPlus PhyTip® Columns, Biotage
Catalog # MSPP-PTB922007
Supplier:  BIOTAGE LLC MS
Supplier Number:  PTB-92-20-07
Specifications
  • Capacity:
    200 µl
  • Description:
    ProPlus PhyTip® columns containing 20 µl affinity media per column, Beckman; Includes buffer kit
  • Cat. No.:
    MSPP-PTB922007
Specifications
About this item
ProPlus PhyTip columns are used for effective automated purification of IgG antibodies.
  • High performance
  • Minimum dilution
  • High throughput
  • Small volume samples
  • Scalable
  • Reproducible
ProPlus (MabSelect Sure™, Cytiva) and ProPlus LX (MabSelect™ SuRe™ LX, Cytiva) are affinity medias with an engineered version of the Protein A affinity ligand that demonstrate attributes of both Protein A and Protein G. The MabSelect™ SuRe™ medias favor an oriented binding of IgGs for high capacity binding of IgGs from low concentration samples and increased selectivity to mouse IgGs. In some cases, MabSelect™ SuRe™ demonstrates a 3-5 fold greater recovery compared to Protein G.
The optimal resin bed volume of the column depends on the total amount of protein present in your samples. If you are working with very low protein concentrations of 10 μg/ml or lower, we recommend using a resin bed volume one or two sizes bigger than indicated below for an effective purification process.
PhyTip columns purify samples by repeated back-and-forth cycles - a process called dual flow chromatography which allows complete binding interactions in each purification step.
Due to maximization of binding of the sample, less protein is lost during the purification process. The result is very high sample recoveries. The unique column design with a packed resin bed results in binding of the entire sample to a small resin bed volume. The purified sample can be eluted in small volumes, resulting in a highly concentrated sample.
Effective purification by PhyTip columns enables a full purification protocol of 96 samples in as little as 15 minutes. Protein A PhyTip columns comes with dual flow chromatography protocols that have been validated with real protein samples and can be transferred to any automated liquid handling system.