ContactUsLinkComponent
DY647 CEP (DyLight 647 phosphoramidite), AAT Bioquest
DY647 CEP (DyLight 647 phosphoramidite), AAT Bioquest
Catalog # 76644-670
Supplier:  AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
DY647 CEP (DyLight 647 phosphoramidite), AAT Bioquest
Catalog # 76644-670
Supplier:  AAT BIOQUEST INC
Supplier Number:  6076
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    For efficiently labeling an oligonucleotide
  • Size:
    100 nmol
  • Cat. No.:
    76644-670
Specifications
About this item
DY647 CEP (DyLight 647 phosphoramidite) is an excellent replacement to the commonly used Cy5 phosphoramidite.
  • High stability
  • Oligo deprotection can be carried out at room temperature
DY647 CEP appears to be more stable than Cy5 phosphoramidite. As Cy3 and Cy5 phosphoramidites, the deprotection of oligos containing DY647 CEP may be carried out with ammonium hydroxide at room temperature, regardless of the base protecting groups on the monomers used.
It is always prudent to use monomers with base labile protecting groups to limit the exposure time to 2 hours or less at 65 °C during deprotection.
Packaging: Plastic vial.
Caution: For research use only.