Specifications
- Temperature range:−18...−24 °C (Preset...−20 °C)
- Capacity:518 L (18.3 cu.ft.)
- Description:Upright manual defrost freezer with solid door
- Environmentally Preferable:
- Model:UFM19W
- Electrical:115 VAC/60 Hz
- Defrost:Manual defrost
- Door Type:One solid swing door, right hinged
- Shelves/drawers:4 adjustable wire shelves
- Weight:98 kg (215.0 lbs.)
- Exterior W×D×H:77×69×180 cm(30.5×27×70.75")
- Interior W×D×H:64×53×163 cm (25.0×20.88×64.0")
- Cat. No.:76644-648
Specifications
About this item
Manual defrost operation creates a more stable environment for the storage of enzymes, reagents, samples, and other laboratory/clinical materials.
- General purpose freezer designed for the storage of samples, enzymes, reagents, and other laboratory and clinical materials
- Large capacity with spacious 18.3 cu.ft. interior
- Static cooling system with manual defrost to ensure a more stable interior
- Factory-installed lock provides security you can count on
- Cantilevered shelving allows for flexible storage
Accucold's general purpose storage line includes upright manual defrost freezers designed for the storage of certain enzymes, reagents, samples, and other materials used in laboratory and clinical settings that require -20 °C temperatures with more stability than traditional no-frost equipment.
Front-opening freezer with an 18.3 cu.ft. interior storage capacity. Manual defrost operation ensures a more stable interior with none of the warming cycles that can compromise temperature in traditional frost-free cooling systems. The dial thermostat is located inside to prevent accidental temperature adjustments. Cantilevered shelving provides more flexibility in storage, with tight spacing between the pilasters so you can adjust as needed. The unit has a flat door liner to accommodate deeper trays. An automatic LED light makes it easy to see the full interior.
This freezer has a white exterior finish. The keyed lock is conveniently located towards the top of the unit for easier user access. The reversible door includes an easy-grip handle in a brushed aluminum finish. This unit includes a 9'5'' long power cord with a hospital grade 'green dot' plug for a more secure connection, ideal for high traffic environments. The cooling system utilizes hydrocarbon refrigerant for a greener footprint. For advanced insulation, the door features a vacuum seal that keeps all cold air inside.
Meets NSF-7 commercial standards and is ETL listed for the U.S. and Canada. It can be customized with additional features to best meet your storage needs, including digital data loggers (DDLs), temperature alarms, NIST calibrated thermometers, and more.
In addition to this freezer, Accucold manufactures frost-free choices for vaccine and laboratory storage, as well as purpose-built refrigeration lines approved to the NSF/ANSI 456 vaccine storage standard.