Betula pendula Recombinant BETV4 Protein (His Tag) (from E. coli)
76631-666
:
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Betula pendula
- Size:250 µg
- Environmentally Preferable:
- Storage Conditions:–20…–80 °C
- Endotoxin Content:Please contact us for more information.
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:BETV4
- Protein/Peptide Name:BETV4
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:The recombinant Betula pendula BETV4 consists of 91 amino acids and predicts a molecular mass of 10.3 kDa.
- Shipping Temperature:Ambient
- Cat. No.:76631-666