Influenza A H1N1 (A/Wisconsin/588/2019)/(A/Victoria/2570/2019) Recombinant Hemagglutinin/HA Protein (His Tag) (from HEK293 Cells)
Catalog # 76632-496
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:H1N1
- Size:100 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg protein as determined by the LAL method.
- Gene ID:EPI1661758 / EPI1799581
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:Hemagglutinin
- Protein/Peptide Name:HA
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:The recombinant hemagglutinin of the Influenza A virus ((A/Wisconsin/588/2019)/(A/Victoria/2570/2019) (H1N1)) consists of 524 amino acids and predicts a molecular mass of 59.2 kDa. It migrates as an approximately 70.8 kDa band in SDS-PAGE under reducing conditions.
- Sequence:Met1-Ile530
- Shipping Temperature:Ambient
- Cat. No.:76632-496
Specifications
About this item
A DNA sequence encoding the Influenza A virus ((A/Wisconsin/588/2019) / (A/Victoria/2570/2019) (H1N1)) hemagglutinin (translated amino acids of EPI1661758 / EPI1799581) (Met1-Ile530), termed as HA, was expressed with a polyhistidine tag at the C-terminus.