Influenza A H1N1 (A/Wisconsin/588/2019)/(A/Victoria/2570/2019) Recombinant Nucleoprotein/NP Protein (His Tag) (from Baculovirus Insect Cells)
76631-864
:
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Baculovirus (Insect cells)
- Species:H1N1
- Size:100 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg protein as determined by the LAL method.
- Gene ID:EPI1661751 / EPI1799574
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:Nucleoprotein
- Protein/Peptide Name:NP
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:The recombinant nucleoprotein of Influenza A virus ((A/Wisconsin/588/2019)/(A/Victoria/2570/2019) (H1N1)) consists of 509 amino acids and predicts a molecular mass of 57.5 kDa.
- Sequence:Met1-Asn498
- Shipping Temperature:Ambient
- Cat. No.:76631-864
A DNA sequence encoding the Influenza A virus ((A/Wisconsin/588/2019) / (A/Victoria/2570/2019) (H1N1)) nucleoprotein (translated amino acids of EPI1661751 / EPI1799574) (Met1-Asn498) was expressed with a polyhistidine tag at the C-terminus.