SARS-CoV (Isolate Tor2) Recombinant Spike RBD Protein (His & AVI Tag), Biotinylated (from HEK293 Cells)
76630-992
:
- Conjugation:Biotin
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:SARS
- Size:20 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg protein as determined by the LAL method.
- Gene ID:NP_828851.1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:ncov RBD|ncov s2|ncov spike|coronavirus s2|S|s1|Spike RBD|ncov s1|coronavirus spike|coronavirus s1|cov spike|RBD
- Protein/Peptide Name:Spike
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:The recombinant SARS-CoV (isolate: Tor2) spike RBD consists of 249 amino acids and predicts a molecular mass of 28.4 kDa. As a result of glycosylation, it migrates as an approximately 37.1 kDa band in SDS-PAGE under reducing conditions.
- Sequence:Arg306-Phe527
- Shipping Temperature:Ambient
- Cat. No.:76630-992
A DNA sequence encoding the SARS-CoV (isolate Tor2) spike RBD (NP_828851.1) (Arg306-Phe527) was expressed with a c-terminal AVI tagged polyhistidine tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.