SARS-CoV (Isolate Tor2) Recombinant Spike S1+S2 (S577A) Protein (ECD, His Tag), Biotinylated (from Baculovirus Insect Cells)
Catalog # 76631-432
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Biotin
- Protein/Peptide Type:Recombinant
- Source:Baculovirus (Insect cells)
- Species:SARS
- Size:20 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg protein as determined by the LAL method.
- Gene ID:NP_828851.1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:ncov RBD|ncov s2|ncov spike|coronavirus s2|S|s1|Spike RBD|ncov s1|coronavirus spike|coronavirus s1|cov spike|RBD
- Protein/Peptide Name:Spike
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:The recombinant SARS-CoV (isolate Tor2) spike S1+S2 consists of 1193 amino acids and predicts a molecular mass of 132.58 kDa. It migrates as an approximately 120.5 kDa band in SDS-PAGE under reducing conditions.
- Sequence:Met1-Pro1195
- Shipping Temperature:Ambient
- Cat. No.:76631-432
Specifications
About this item
A DNA sequence encoding the full-lenght ECD of SARS-CoV (isolate Tor2) spike (NP_828851.1) (Met1-Pro1195) was expressed with a C-terminal polyhistidine tag. The purified protein was biotinylated in vitro.