SARS-CoV-2 (2019-nCoV) Recombinant Spike RBD(Q414E)-His Protein (from HEK293 Cells)
76631-142
:
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:SARS-CoV-2
- Size:100 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg protein as determined by the LAL method.
- Gene ID:YP_009724390.1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:ncov RBD|ncov s2|ncov spike|coronavirus s2|S|s1|Spike RBD|ncov s1|coronavirus spike|coronavirus s1|cov spike|RBD
- Protein/Peptide Name:Spike
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:The recombinant SARS-CoV-2 (2019-nCoV) Spike RBD(Q414E)-His Recombinant Protein consists of 234 amino acids and predicts a molecular mass of 26.5 kDa.
- Sequence:Arg319-Phe541(Q414E)
- Shipping Temperature:Ambient
- Cat. No.:76631-142
A DNA sequence encoding the SARS-CoV-2 (2019-nCoV) Spike RBD(Q414E)-His Recombinant Protein (YP_009724390.1) (Arg319-Phe541(Q414E)) was expressed with a polyhistidine tag at the C-terminus.