Chinese hamster Recombinant Clusterin Protein (His Tag) (from CHO cells)
A DNA sequence encoding the chinese hamster CLU (G3HNJ3-1) (Glu22-Glu447) was expressed with a polyhistidine tag at the C-terminus.
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:CHO cells
- Species:Chinese hamster
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Gene ID:G3HNJ3-1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:SP-40|D14Ucla3|AI893575|Sgp-2|Cli|ApoJ|Sgp2|Sugp-2
- Protein/Peptide Name:CLU
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:The recombinant chinese hamster CLU consists of 437 amino acids and predicts a molecular mass of 50.9 kDa. It migrates as an approximately 63.5, 38.6 and 29.3 kDa in SDS-PAGE under reducing conditions.
- Sequence:Met1-Glu447
- Shipping Temperature:Ambient
Frequently Bought Together