Order Entry
United States
ContactUsLinkComponent
SARS-CoV-2 Recombinant Spike S1+S2 (D614G) trimer Protein (ECD, His tag) (HPLC-verified) (from HEK293 Cells)
SARS-CoV-2 Recombinant Spike S1+S2 (D614G) trimer Protein (ECD, His tag) (HPLC-verified) (from HEK293 Cells)
Catalog # 76631-330
Supplier:  Sino Biological
CAS Number:  
SARS-CoV-2 Recombinant Spike S1+S2 (D614G) trimer Protein (ECD, His tag) (HPLC-verified) (from HEK293 Cells)
Catalog # 76631-330
Supplier:  Sino Biological
Supplier Number:  40589-V08H8-100
CAS Number:  

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    SARS-CoV-2
  • Size:
    100 µg
  • Environmentally Preferable:
  • Storage Conditions:
    –20…–80 °C
  • Endotoxin Content:
    <1.0 EU per μg of the protein as determined by the LAL method
  • Biological Activity:
    Immobilized human ACE2 protein (mFc tag)(Cat:10108-H05H) at 2 μg/ml (100 μl/well) can bind SARS-CoV-2 Spike S1+S2 (D614G) trimer Protein (ECD, His tag)(Cat:40589-V08H8),the EC50 of SARS-CoV-2 Spike S1+S2 (D614G) trimer Protein (ECD, His tag)(Cat:40589-V08H8) is 4-24 ng/ml.
  • Gene ID:
    YP_009724390.1
  • Reconstitution Instructions:
    A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
  • Carrier-Free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    ncov RBD|ncov s2|ncov spike|coronavirus s2|S|s1|Spike RBD|ncov s1|coronavirus spike|coronavirus s1|cov spike|RBD
  • Protein/Peptide Name:
    Spike
  • Purity:
    >90% as determined by SDS-PAGE.
    >90% as determined by SEC-HPLC.
  • Molecular Weight:
    The recombinant SARS-CoV-2 Spike S1+S2 consists of 1231 amino acids and predicts a molecular mass of 136.47 kDa.
  • Shipping Temperature:
    Ambient
  • Cat. No.:
    76631-330

Specifications

About this item

A DNA sequence encoding the SARS-CoV-2 Spike S1+S2 (YP_009724390.1, with mutations D614G, F817P, A892P, A899P, A942P, K986P, V987P and furin cleavage site mutants) expressed with the bacteriophage T4 fibritin and a polyhistidine tag at the C-terminus.