SARS-CoV-2 (2019-nCoV) Recombinant Spike S1+S2 ECD (HV69-70 deletion, Y144 deletion, N501Y, A570D, D614G, P681H, T716I, S982A, D1118H)-His Recombinant Protein (from Baculovirus Insect cells)
Catalog # 76631-582
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Baculovirus (Insect cells)
- Species:SARS-CoV-2
- Size:100 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Biological Activity:Measured by its binding ability in a functional ELISA. Immobilized Human ACE2 (mFc tag)(Cat: 10108-H05H) at 2 μg/ml (100 μl/well) can bind SARS-CoV-2 (2019-nCoV) Spike S1+S2 ECD (HV69-70 deletion, Y144 deletion, N501Y, A570D, D614G, P681H, T716I, S982A, D1118H)-His(Cat:40589-V08B6), the EC50 of SARS-CoV-2 (2019-nCoV) Spike S1+S2 ECD (HV69-70 deletion, Y144 deletion, N501Y, A570D, D614G, P681H, T716I, S982A, D1118H)-His(Cat:40589-V08B6) is 200-800 ng/ml.
- Gene ID:YP_009724390.1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:ncov RBD|ncov s2|ncov spike|coronavirus s2|S|s1|Spike RBD|ncov s1|coronavirus spike|coronavirus s1|cov spike|RBD
- Protein/Peptide Name:Spike
- Purity:>85% as determined by SDS-PAGE
- Molecular Weight:The recombinant SARS-CoV-2 (2019-nCoV) Spike Protein (S1+S2 ECD(HV69-70 deletion, Y144 deletion, N501Y, A570D, D614G, P681H, T716I, S982A, D1118H), His tag) consists of 1206 amino acids and predicts a molecular mass of 134.1 kDa.
- Sequence:Met1-Pro1213(del(HV69-70 deletion, Y144 deletion, N501Y, A570D, D614G, P681H, T716I, S982A, D1118H)
- Shipping Temperature:Ambient
- Cat. No.:76631-582
Specifications
About this item
A DNA sequence encoding the SARS-CoV-2 (2019-nCoV) Spike Protein (S1+S2 ECD) (YP_009724390.1) (Met1-Pro1213(HV69-70 deletion, Y144 deletion, N501Y, A570D, D614G, P681H, T716I, S982A, D1118H)) was expressed with a polyhistidine tag at the C-terminus.