4-Hydroxy-2,2,6,6-tetramethylpiperidinooxy (4-hydroxy-TEMPO, TEMPOL)
Catalog # 80503-466
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Plastic ampoule
- Size:500 mg
- Cat. No.:80503-466
- Supplier no.:581500-500MG
Specifications
About this item
:
:172.25 g/mol
CAS Number:2226-96-2
Formula:C₉H₁₈NO₂
MDL Number:MFCD00006478