Ammonium sulfate
Catalog # 80057-798
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:1 kg
- Environmentally Preferable:
- Cat. No.:80057-798
- Supplier no.:168355-1KG
Specifications
About this item
Synonyms: sulfuric acid diammonium salt, Diammonium sulfate, Sulphuric acid diammonium salt, Diammonium sulphate, Mascagnite
:
:132.14 g/mol
CAS Number:7783-20-2
Formula:(NH₄)₂SO₄
MDL Number:MFCD00003391
Merck Index:13,00559