TI-Innovator Vibration Motor Pack
Catalog # 470348-860
Supplier: D&H
Specifications
- Description:TI-Innovator vibration motor module, 5 pack
- Cat. No.:470348-860
Specifications
About this item
Add motion/sound to your TI-Innovator hub system.
- Practice programming skills by physically controlling a continuous rotation servo
- Students see the physical results of their code
These coin-type motors vibrate like your smart phone. Each one comes mounted on a printed circuit board for easy handling, with a cable to connect it to the TI-Innovator hub for programming. Build and program your Internet of Things (IoT) in your classroom using the TI-Innovator hub system and your existing TI-Nspire or TI-84 Plus CE technology.
Delivery: Pack includes five vibration motors and cables.