ContactUsLinkComponent
TI-Innovator Moisture Module
TI-Innovator Moisture Module
Catalog # 470348-846
Supplier:  D&H
undefined
TI-Innovator Moisture Module
Catalog # 470348-846
Supplier:  D&H
Supplier Number:  MOISTMOD5PK
Specifications
  • Description:
    TI-Innovator moisture module, 5 pack
  • Cat. No.:
    470348-846
Specifications
About this item
Soil moisture sensor for your TI-Innovator hub system.
  • Collect soil moisture data as inputs for programming
  • Real-world applications for your TI-Innovator hub
The TI-Innovator moisture sensor detects the moisture of soil, to judge if there is dampness around the sensor. It comes mounted on a printed circuit board for easy handling, with a cable to connect it to the TI-Innovator hub for programming. Build and program your Internet of Things (IoT) in your classroom using the TI-Innovator hub system and your existing TI-Nspire or TI-84 Plus CE technology.
Delivery: Pack includes five moisture sensors and cables.