Orders LinkContactUsLinkComponent
TI-SensorLink Adapter
TI-SensorLink Adapter
TI-Innovator SensorLink adapter
Catalog # 470348-866
Supplier:   D&H
Supplier #:  SENSORLINKADAPT
TI-SensorLink Adapter
Catalog # 470348-866
Supplier:  D&H
TI-SensorLink Adapter
TI-SensorLink Adapter
TI-Innovator SensorLink adapter
Price
$16.99
Log in for availability
Each
Product Details & Documents
Integrate your vernier BTA sensors with your TI-Innovator hub system.
  • Extend the utility and capability of your TI-Innovator hub using wired vernier sensors
  • Diversify the types of inputs that can provide data for programming
The TI-SensorLink adapter allows for connection from several popular vernier sensors to the TI-Innovator hub. Use the pH, temperature, force, pressure, low-g accelerometer, light and energy sensors from vernier in your TI-Innovator activities.
Specifications
  • Description:
    TI-Innovator SensorLink adapter