Orders LinkContactUsLinkComponent
TI-RGB Array
TI-RGB Array
TI-RGB array
Catalog # 470348-868
Supplier:   D&H
Supplier #:  TIRGBARRAY
TI-RGB Array
Catalog # 470348-868
Supplier:  D&H
TI-RGB Array
TI-RGB Array
TI-RGB array
Price
$16.99
Log in for availability
Each
Product Details & Documents
Engage students with this colorful output device.
  • Practice programming skills by controlling LED array
  • Students see the physical results of their code
The TI-RBG array contains 16 individually programmable RGB LEDs for use with the TI-Innovator hub system.
Delivery information: Package includes one TI-RGB array and cable.
Specifications
  • Description:
    TI-RGB array
  • Cat. No.:
    470348-868