ContactUsLinkComponent
Vernier® Optical DO Probe Metal Guard
  470219-314
undefined
Vernier® Optical DO Probe Metal Guard
  470219-314
 :  ODO-GRD
  • Description:
    Optical DO Probe Metal Guard
  • Cat. No.:
    470219-314
  • Supplier no.:
    ODO-GRD
The Optical DO Probe Metal Guard protects the Vernier Optical DO Probe and the Go Direct Optical DO Probe cap when taking measurements in the field and helps weigh down the probe when submerged.
Educational use only: Vernier products are designed for educational use. They are not appropriate for industrial, medical, or commercial applications.