C-Slide Chamber Slide, NanoEnTek
Catalog # 76627-758
Supplier: NANOENTEK USA INC.
Specifications
- Description:C-Slide cell counting chamber slide
- Dimensions:25×75 mm
- Cat. No.:76627-758
- No. of chambers:2
- Chamber size:10 µl
Specifications
About this item
C-Slide is a disposable cell counting chamber slide compatible with all Countess series, as well as EVE/EVE Plus cell counters.
- For use with automated cell counters
- No need for coverslips or washing (disposable slides)
- Staining method: Trypan Blue
- Reduce exposure to infectious samples and contamination
- Reproducibility and reliability guaranteed
- Made of sturdy quartz grade optical plastic
C-Slide is the same as Countess Slide. NanoEnTek is an original developer and manufacturer of the Countess and Countess Slide. C-Slide consists of two separate enclosed chambers each with two ports for sample injection. Each C-Slide can measure two different samples or two of the same sample.
Unlike a conventional hemocytometer, C-Slide is completely disposable and packed individually, hence convenient and contamination-free.