CR1500 Handheld Wired Barcode Scanner and Barcode Scanner with Stand; 1D, 2D, QR Code, Brady Worldwide
Save valuable time with a fast and reliable scanning solution. This omnidirectional barcode reader eliminates repositioning and time consuming re-scans of 1D and 2D barcodes that have seen challenging conditions.
- Compact, lightweight and rugged to survive the toughest industrial environments
- Fast and easy scanning with a high-performance scan engine and intuitive targeting
- Read both high-density and wide-field barcodes with a single device via dual-field optics
Smart scanning in a small package, the CR1500 handheld wired barcode scanner is one of the smallest barcode readers on the market. The PVC-free housing is IP 54 rated to protect against harsh chemical cleaning agents, dust, and water ingress.
Frequently Bought Together