EP Reference Solution, Red Reference Solution, RICCA Chemical Company
About this item
12 Options Available Below
Product Details & Documents
Certificates
Search by Lot NumberSpecifications for Product Family
Specifications
Specifications
- Catalog No:
- RCR2882200120C
- RCR2882200-1C
- RCR2882300120C
- RCR2882300-1C
- RCR2882400120C
- RCR2882400-1C
- RCR2882500120C
- RCR2882500-1C
- RCR2882600120C
- RCR2882600-1C
- RCR2882700120C
- RCR2882700-1C
- Appearance:
- Red liquid
- Red liquid
- Red liquid
- Red liquid
- Red liquid
- Red liquid
- Red liquid
- Red liquid
- Red liquid
- Red liquid
- Red liquid
- Red liquid
- Storage Temperature:
- Ambient
- Ambient
- Ambient
- Ambient
- Ambient
- Ambient
- Ambient
- Ambient
- Ambient
- Ambient
- Ambient
- Ambient
Specifications
Product Family Options
Product Information
- SizeAvailabilityPrice
- RCR2882200120CEP Reference Solution R2, Red Reference SolutionEP Reference Solution R2, Red Reference Solution120 mLLog in for availabilityEach(1/Itm/Bx)$111.55Supplier Number: R2882200-120CItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.RCR2882200120CAppearanceRed liquidStorage TemperatureAmbient - RCR2882200-1CEP Reference Solution R2, Red Reference SolutionSupplier Number: R2882200-1CSpecifications:
More
Specifications
Cat. No.RCR2882200-1CAppearanceRed liquidStorage TemperatureAmbient - RCR2882300120CEP Reference Solution R3, Red Reference SolutionEP Reference Solution R3, Red Reference Solution120 mLLog in for availabilityEach(1/Itm/Bx)$105.34Supplier Number: R2882300-120CSpecifications:
More
Specifications
Cat. No.RCR2882300120CAppearanceRed liquidStorage TemperatureAmbient - RCR2882300-1CEP Reference Solution R3, Red Reference SolutionSupplier Number: R2882300-1CSpecifications:
More
Specifications
Cat. No.RCR2882300-1CAppearanceRed liquidStorage TemperatureAmbient - RCR2882400120CEP Reference Solution R4, Red Reference SolutionEP Reference Solution R4, Red Reference Solution120 mLLog in for availabilityEach(1/Itm/Bx)$112.60Supplier Number: R2882400-120CItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.RCR2882400120CAppearanceRed liquidStorage TemperatureAmbient - RCR2882400-1CEP Reference Solution R4, Red Reference SolutionSupplier Number: R2882400-1CSpecifications:
More
Specifications
Cat. No.RCR2882400-1CAppearanceRed liquidStorage TemperatureAmbient - RCR2882500120CEP Reference Solution R5, Red Reference SolutionSupplier Number: R2882500-120CItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.RCR2882500120CAppearanceRed liquidStorage TemperatureAmbient - RCR2882500-1CEP Reference Solution R5, Red Reference SolutionSupplier Number: R2882500-1CSpecifications:
More
Specifications
Cat. No.RCR2882500-1CAppearanceRed liquidStorage TemperatureAmbient - RCR2882600120CEP Reference Solution R6, Red Reference SolutionEP Reference Solution R6, Red Reference Solution120 mLLog in for availabilityEach(1/Itm/Bx)$103.28Supplier Number: R2882600-120CItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.RCR2882600120CAppearanceRed liquidStorage TemperatureAmbient - RCR2882600-1CEP Reference Solution R6, Red Reference SolutionSupplier Number: R2882600-1CSpecifications:
More
Specifications
Cat. No.RCR2882600-1CAppearanceRed liquidStorage TemperatureAmbient - RCR2882700120CEP Reference Solution R7, Red Reference SolutionEP Reference Solution R7, Red Reference Solution120 mLLog in for availabilityEach(1/Itm/Bx)$112.59Supplier Number: R2882700-120CItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.RCR2882700120CAppearanceRed liquidStorage TemperatureAmbient - RCR2882700-1CEP Reference Solution R7, Red Reference SolutionSupplier Number: R2882700-1CSpecifications:
More
Specifications
Cat. No.RCR2882700-1CAppearanceRed liquidStorage TemperatureAmbient
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



