Order Entry
United States
Orders LinkContactUsLinkComponent
 

EP Reference Solution, Brown Reference Solution, RICCA Chemical Company

EP Reference Solution, Brown Reference Solution, RICCA Chemical Company
Supplier:  Ricca Chemical
Certificates
EP Reference Solution, Brown Reference Solution, RICCA Chemical Company
EP Reference Solution, Brown Reference Solution, RICCA Chemical Company

12 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

 

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • RCR2880100120C
  • RCR2880100-1C
  • RCR2880200120C
  • RCR2880200-1C
  • RCR2880300120C
  • RCR2880300-1C
  • RCR2880400-1C
  • RCR2880500120C
  • RCR2880500-1C
  • RCR2880700-1C
  • RCR2880800-1C
  • RCR2880900-1C
  • Appearance:
  • Brown liquid
  • Brown liquid
  • Brown liquid
  • Brown liquid
  • Brown liquid
  • Brown liquid
  • Brown liquid
  • Brown liquid
  • Brown liquid
  • Brown liquid
  • Brown liquid
  • Brown liquid
  • Storage Temperature:
  • Ambient
  • Ambient
  • Ambient
  • Ambient
  • Ambient
  • Ambient
  • Ambient
  • Ambient
  • Ambient
  • Ambient
  • Ambient
  • Ambient

Specifications

Product Family Options

Product Information

  • Size
    Availability
    Price
  • RCR2880100120C
    EP Reference Solution B1, Brown Reference Solution
    EP Reference Solution B1, Brown Reference Solution
    120 mL
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $112.60
    $112.60
    Each(1/Itm/Bx)
       $112.60

    Supplier Number: R2880100-120C
    Specifications:
    More
    Specifications
    Cat. No.
    RCR2880100120C
    Appearance
    Brown liquid
    Storage Temperature
    Ambient
  • RCR2880100-1C
    EP Reference Solution B1, Brown Reference Solution
    EP Reference Solution B1, Brown Reference Solution
    1 L
    Log in for availability
    Log in for availability
    Each(1l)
    Each(1l) - $423.53
    $423.53
    Each(1l)
       $423.53

    Supplier Number: R2880100-1C
    Specifications:
    More
    Specifications
    Cat. No.
    RCR2880100-1C
    Appearance
    Brown liquid
    Storage Temperature
    Ambient
  • RCR2880200120C
    EP Reference Solution B2, Brown Reference Solution
    EP Reference Solution B2, Brown Reference Solution
    120 mL
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $119.82
    $119.82
    Each(1/Itm/Bx)
       $119.82

    Supplier Number: R2880200-120C
    Specifications:
    More
    Specifications
    Cat. No.
    RCR2880200120C
    Appearance
    Brown liquid
    Storage Temperature
    Ambient
  • RCR2880200-1C
    EP Reference Solution B2, Brown Reference Solution
    EP Reference Solution B2, Brown Reference Solution
    1 L
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $501.00
    $501.00
    Each(1/Itm/Bx)
       $501.00

    Supplier Number: R2880200-1C
    Specifications:
    More
    Specifications
    Cat. No.
    RCR2880200-1C
    Appearance
    Brown liquid
    Storage Temperature
    Ambient
  • RCR2880300120C
    EP Reference Solution B3, Brown Reference Solution
    EP Reference Solution B3, Brown Reference Solution
    120 mL
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $97.10
    $97.10
    Each(1/Itm/Bx)
       $97.10

    Supplier Number: R2880300-120C
    Specifications:
    More
    Specifications
    Cat. No.
    RCR2880300120C
    Appearance
    Brown liquid
    Storage Temperature
    Ambient
  • RCR2880300-1C
    EP Reference Solution B3, Brown Reference Solution
    EP Reference Solution B3, Brown Reference Solution
    1 L
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $364.66
    $364.66
    Each(1/Itm/Bx)
       $364.66

    Supplier Number: R2880300-1C
    Specifications:
    More
    Specifications
    Cat. No.
    RCR2880300-1C
    Appearance
    Brown liquid
    Storage Temperature
    Ambient
  • RCR2880400-1C
    EP Reference Solution B4, Brown Reference Solution
    EP Reference Solution B4, Brown Reference Solution
    1 L
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $435.91
    $435.91
    Each(1/Itm/Bx)
       $435.91

    Supplier Number: R2880400-1C
    Specifications:
    More
    Specifications
    Cat. No.
    RCR2880400-1C
    Appearance
    Brown liquid
    Storage Temperature
    Ambient
  • RCR2880500120C
    EP Reference Solution B5, Brown Reference Solution
    EP Reference Solution B5, Brown Reference Solution
    120 mL
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $119.84
    $119.84
    Each(1/Itm/Bx)
       $119.84

    Supplier Number: R2880500-120C
    Specifications:
    More
    Specifications
    Cat. No.
    RCR2880500120C
    Appearance
    Brown liquid
    Storage Temperature
    Ambient
  • RCR2880500-1C
    EP Reference Solution B5, Brown Reference Solution
    EP Reference Solution B5, Brown Reference Solution
    1 L
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $511.34
    $511.34
    Each(1/Itm/Bx)
       $511.34

    Supplier Number: R2880500-1C
    Specifications:
    More
    Specifications
    Cat. No.
    RCR2880500-1C
    Appearance
    Brown liquid
    Storage Temperature
    Ambient
  • RCR2880700-1C
    EP Reference Solution B7, Brown Reference Solution
    EP Reference Solution B7, Brown Reference Solution
    1 L
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $316.10
    $316.10
    Each(1/Itm/Bx)
       $316.10

    Supplier Number: R2880700-1C
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    RCR2880700-1C
    Appearance
    Brown liquid
    Storage Temperature
    Ambient
  • RCR2880800-1C
    EP Reference Solution B8, Brown Reference Solution
    EP Reference Solution B8, Brown Reference Solution
    1 L
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $391.50
    $391.50
    Each(1/Itm/Bx)
       $391.50

    Supplier Number: R2880800-1C
    Specifications:
    More
    Specifications
    Cat. No.
    RCR2880800-1C
    Appearance
    Brown liquid
    Storage Temperature
    Ambient
  • RCR2880900-1C
    EP Reference Solution B9, Brown Reference Solution
    EP Reference Solution B9, Brown Reference Solution
    1 L
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $295.43
    $295.43
    Each(1/Itm/Bx)
       $295.43

    Supplier Number: R2880900-1C
    Specifications:
    More
    Specifications
    Cat. No.
    RCR2880900-1C
    Appearance
    Brown liquid
    Storage Temperature
    Ambient

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.