Order Entry
United States
ContactUsLinkComponent
EP Reference Solution, Greenish-Yellow Reference Solution, RICCA Chemical Company
EP Reference Solution, Greenish-Yellow Reference Solution, RICCA Chemical Company
Supplier:  Ricca Chemical
Certificates
undefined
EP Reference Solution, Greenish-Yellow Reference Solution, RICCA Chemical Company
Supplier:  Ricca Chemical
Certificates

Specifications

  • Storage Temperature:
    Ambient
  • Appearance:
    Greenish-yellow liquid

Specifications

Related Information

Frequently Bought Together