EP Reference Solution, Yellow Reference Solution, RICCA Chemical Company
Supplier: Ricca Chemical
EP Reference Solution, Yellow Reference Solution, RICCA Chemical Company
Supplier: Ricca Chemical
Certificates
About this item
9 Options Available Below
Product Details & Documents
Documents
Certificates
Search by Lot NumberSpecifications for Product Family
Specifications
Specifications
- Catalog No:
- RCR2883100-1C
- RCR2883200120C
- RCR2883200-1C
- RCR2883300120C
- RCR2883300-1C
- RCR2883400-1C
- RCR2883500-1C
- RCR2883600-1C
- RCR2883700-1C
- Appearance:
- Yellow liquid
- Yellow liquid
- Yellow liquid
- Yellow liquid
- Yellow liquid
- Yellow liquid
- Yellow liquid
- Yellow liquid
- Yellow liquid
- Storage Temperature:
- Ambient
- Ambient
- Ambient
- Ambient
- Ambient
- Ambient
- Ambient
- Ambient
- Ambient
Specifications
Product Family Options
Product Information
- SizeAvailabilityPrice
- RCR2883100-1CEP Reference Solution Y1, Yellow Reference SolutionEP Reference Solution Y1, Yellow Reference Solution1 LLog in for availabilityEach(1/Itm/Bx)$258.24Supplier Number: R2883100-1CSpecifications:
More
Specifications
Cat. No.RCR2883100-1CAppearanceYellow liquidStorage TemperatureAmbient - RCR2883200120CEP Reference Solution Y2, Yellow Reference SolutionEP Reference Solution Y2, Yellow Reference Solution120 mLLog in for availabilityEach(1/Itm/Bx)$96.07Supplier Number: R2883200-120CSpecifications:
More
Specifications
Cat. No.RCR2883200120CAppearanceYellow liquidStorage TemperatureAmbient - RCR2883200-1CEP Reference Solution Y2, Yellow Reference SolutionEP Reference Solution Y2, Yellow Reference Solution1 LLog in for availabilityEach(1/Itm/Bx)$324.37Supplier Number: R2883200-1CSpecifications:
More
Specifications
Cat. No.RCR2883200-1CAppearanceYellow liquidStorage TemperatureAmbient - RCR2883300120CEP Reference Solution Y3, Yellow Reference SolutionEP Reference Solution Y3, Yellow Reference Solution120 mLLog in for availabilityEach(1/Itm/Bx)$92.98Supplier Number: R2883300-120CSpecifications:
More
Specifications
Cat. No.RCR2883300120CAppearanceYellow liquidStorage TemperatureAmbient - RCR2883300-1CEP Reference Solution Y3, Yellow Reference SolutionEP Reference Solution Y3, Yellow Reference Solution1 LLog in for availabilityEach(1/Itm/Bx)$297.50Supplier Number: R2883300-1CSpecifications:
More
Specifications
Cat. No.RCR2883300-1CAppearanceYellow liquidStorage TemperatureAmbient - RCR2883400-1CEP Reference Solution Y4, Yellow Reference SolutionEP Reference Solution Y4, Yellow Reference Solution1 LLog in for availabilityEach(1/Itm/Bx)$248.94Supplier Number: R2883400-1CSpecifications:
More
Specifications
Cat. No.RCR2883400-1CAppearanceYellow liquidStorage TemperatureAmbient - RCR2883500-1CEP Reference Solution Y5, Yellow Reference SolutionEP Reference Solution Y5, Yellow Reference Solution1 LLog in for availabilityEach(1/Itm/Bx)$346.04Supplier Number: R2883500-1CSpecifications:
More
Specifications
Cat. No.RCR2883500-1CAppearanceYellow liquidStorage TemperatureAmbient - RCR2883600-1CEP Reference Solution Y6, Yellow Reference SolutionEP Reference Solution Y6, Yellow Reference Solution1 LLog in for availabilityEach(1/Itm/Bx)$293.37Supplier Number: R2883600-1CSpecifications:
More
Specifications
Cat. No.RCR2883600-1CAppearanceYellow liquidStorage TemperatureAmbient - RCR2883700-1CEP Reference Solution Y7, Yellow Reference SolutionEP Reference Solution Y7, Yellow Reference Solution1 LLog in for availabilityEach(1/Itm/Bx)$236.56Supplier Number: R2883700-1CSpecifications:
More
Specifications
Cat. No.RCR2883700-1CAppearanceYellow liquidStorage TemperatureAmbient
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



