Lockout Station with Keyed Alike Nylon Safety Lockout Padlocks, Brady Worldwide
Catalog # 76637-572
Supplier: Brady Worldwide
Specifications
- Color:Red
- Description:Lockout Kit, keyed different
- Cat. No.:76637-572
Specifications
About this item
Safely power down faster when powering up lockout tagout readiness with this convenient, easily accessible and portable forklift lockout kit.
- OSHA compliance driver
- Built to last
Designed and assembled exclusively for safety to help users comply with OSHA 29 CFR § 1910.147 Control of Hazardous Energy. All components meet Brady’s highest standards for quality, durability and reliability.
BatteryBlock™ Power Connector Lockout device fits on 175A and 350A power connectors with and without handles. Non-conductive pins ensure the device stays secure during maintenance projects and off-shift periods. Lock out more than forklifts; use on pallet jacks, powered carts, scissor lifts, and stackers.
Ordering information: Lockout Kit includes: (1) Nylon Safety Lockout Padlock, (1) BatteryBlock Power Connector Lockout - Forklift, (1) Lockout Tag, and (1) Zip Tie.