HYDRANAL™ - NEXTGEN Coulomat A-FA, Anolyte for Coulometric Karl Fischer Titration, Fluka™
Supplier: Honeywell Research Chemicals
Certificates
About this item
Anolyte for coulometric Karl Fischer titration of ketones and Li-ion battery electrolytes, preferred for cells with diaphragm, acetonitrile-based, free of alcohols (free of CMR substances)
- Assay of water Max. 50 ppm
- Number of samples Min. 2
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.