Eppendorf FAST PCR Tube Strips
Supplier: Eppendorf
Certificates
About this item
Eppendorf Fast PCR tube strips made of polyethylene transfer heat faster than tubes made of traditional plastics. The fast PCR tube strips are particularly suited to scientists using fast PCR reagent kits, scientists trying to speed up protocols or cyclers with high heating and cooling rates.
- Ideal for use with fast cyclers and fast enzymes
- Eight reaction tubes in strip-format – ideal for small sample volumes
- Easily adaptable for automation
- Sealable using flat or domed strip lids
- Certified free from human DNA, DNase, RNase and PCR inhibitors
The fast PCR tube strip allows scientists using fast cyclers (such as the Eppendorf Mastercycler X50) and fast enzymes to obtain higher PCR yield. This is achieved by the faster heat transfer from block to reaction, allowing for better control over the reaction. The tube strips are suitable for fast end-point PCR and real-time PCR applications.
Specifications
- Catalog No:
- 76289-506
- 76289-504
- 76289-508
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental Certification:
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001