Order Entry
United States
ContactUsLinkComponent
 
2D Coded Sample Storage Tubes, 0.26 ml, 96-format, External Thread, Azenta Life Sciences
 
undefined
2D Coded Sample Storage Tubes, 0.26 ml, 96-format, External Thread, Azenta Life Sciences
 

 

  • Catalog No:
  • 76636-632
  • 76637-118
  • 76636-672
  • 76637-102
  • 76636-884
  • 76636-582
  • 76637-010
  • Height with cap:
  • 18.6 mm
  • 18.6 mm
  • 18.6 mm
  • 18.6 mm
  • 18.6 mm
  • 18.6 mm
  • 18.6 mm
  • Ext.Ø with cap:
  • 8.7 mm
  • 8.7 mm
  • 8.7 mm
  • 8.7 mm
  • 8.7 mm
  • 8.7 mm
  • 8.7 mm
  • Centre to centre:
  • 9.0 mm
  • 9.0 mm
  • 9.0 mm
  • 9.0 mm
  • 9.0 mm
  • 9.0 mm
  • 9.0 mm
  • Height in rack:
  • 19.13 mm
  • 19.13 mm
  • 19.13 mm
  • 19.13 mm
  • 19.13 mm
  • 19.13 mm
  • 19.13 mm
  • Overall rack height incl. lid:
  • 22.0 mm
  • 22.0 mm
  • 22.0 mm
  • 22.0 mm
  • 22.0 mm
  • 22.0 mm
  • 22.0 mm

 

Empty racks

 

Frequently Bought Together