Thermal Bond Heat Sealing Foils, Polystyrene, Peelable
The thermal bond heat seal is a heavy duty laminate foil seal suitable for providing a very strong, but peelable seal. Suitable for long term, very low temperate storage.
- Recommended for PCR including with water bath thermal cyclers
- Compatible with polypropylene plates
- Pierceable with blade
- Peelable
- High solvent resistance, including DMSO
- Autoclavable (121 °C)
- Temperature range: –200 to +110 °C
- Non sterile
Heat sealing offers a 100% effective method of plate sealing, for complete seal integrity, as well as being quick and cost effective.
The sheet seals fit all standard SBS footprint PCR and qPCR plates, microplates, assay and storage plates, from 12 to 1536 wells.
: Available as sheets for use with manual and semiautomated sealers. Available in multiple roll formats compatible with specified automated heat sealers. Gamma treatment available upon request.
Frequently Bought Together