Order Entry
United States
ContactUsLinkComponent
2D Coded Sample Storage Tubes, 0.48 ml, 96-format, Internal Thread, Azenta Life Sciences
2D Coded Sample Storage Tubes, 0.48 ml, 96-format, Internal Thread, Azenta Life Sciences
Supplier:  AZENTA US, INC
Certificates
undefined
2D Coded Sample Storage Tubes, 0.48 ml, 96-format, Internal Thread, Azenta Life Sciences
Supplier:  AZENTA US, INC
Certificates

Specifications

  • Catalog No:
  • 76636-758
  • 76637-108
  • 76636-558
  • 76636-984
  • 76636-614
  • 76637-098
  • Height with cap:
  • 34.7 mm
  • 34.7 mm
  • 34.7 mm
  • 34.7 mm
  • 34.7 mm
  • 34.7 mm
  • Ext.Ø with cap:
  • 8.6 mm
  • 8.6 mm
  • 8.6 mm
  • 8.6 mm
  • 8.6 mm
  • 8.6 mm
  • Centre to centre:
  • 9.0 mm
  • 9.0 mm
  • 9.0 mm
  • 9.0 mm
  • 9.0 mm
  • 9.0 mm
  • Height in rack:
  • 36.2 mm
  • 36.2 mm
  • 36.2 mm
  • 36.2 mm
  • 36.2 mm
  • 36.2 mm
  • Overall rack height incl. lid:
  • 44.9 mm
  • 44.9 mm
  • 44.9 mm
  • 44.9 mm
  • 44.9 mm
  • 44.9 mm

Specifications

Empty racks

Product Information

Frequently Bought Together