Storage Microplates, 96 Round Well, Deep Well, for Magnetic Separators, Azenta Life Sciences
Supplier: AZENTA US, INC
Certificates
About this item
These 96 deep well storage microplates are especially designed for use with magnetic separators for bead separation protocols. Clear plate, perfect for storage applications.
- Made from polypropylene, a material with very low biomolecular binding characteristics, a high temperature tolerance and resistance to many standard laboratory chemicals, including DMSO
- Autoclavable and stackable
- Free from RNase, DNase, human genomic DNA and endotoxins
- Alphanumeric grid reference to aid well and sample identification
- SBS footprint
- Round wells, suitable for most applications as they reduce droplet effects and wicking
- Temperature range for use: –80 to 120 °C
- Suitable as a collection plate from filter systems; compatible with plate readers, ideal for use with automation
Perfect fit to magnetic separators: the magnetic beads separate perfectly into e.g. a ring (if a ring magnet is used) allowing easy removal of supernatant from the center of the well by manual or automated pipetting.
Best for small volume recovery due to the V-shaped bottom, as the liquid gathers at the lowest point of the V for easy pipetting.
Ultra-low DNA binding option available (4ti-LB0125) for sensitive applications with ultra-low DNA input and for maximum DNA recovery after low temperature storage and high temperature incubation.
Frequently Bought Together