Adhesive Seal Roller and Applicator
Supplier: AZENTA US, INC
Certificates
About this item
The Adhesive Seal Roller and Adhesive Seal Applicator ensure even pressure is applied across the adhesive seal for a complete application to the plate, across every well.
- The handle of the Adhesive Seal Roller is made of a durable plastic, with a semi-hard padded rolling wheel
- The straight rigid sides of the small-sized Adhesive Seal Applicator allow for even pressure application
When applying adhesive seals to 384 well plates, we recommend using the application tools in conjunction with the FrameStar 384 Holder (4ti-0391) to support the 384 well plates during seal applications, and to give a level base.
Frequently Bought Together