ContactUsLinkComponent
Cap Mat, PCR Plate, Azenta Life Sciences
Cap Mat, PCR Plate, Azenta Life Sciences
Catalog # 76638-012
Supplier:  AZENTA US, INC
Cap Mat, PCR Plate, Azenta Life Sciences
Catalog # 76638-012
Supplier:  AZENTA US, INC
Supplier Number:  4TI-0778
Specifications
  • Optical property:
    Light blocking
  • Sterility:
    Non sterile
  • Description:
    Cap mat for pcr plates, 96 individual caps in sheet format, blue tpe, pierceable
  • Material:
    TPE
  • Cat. No.:
    76638-012
Specifications
About this item
These 96 individual caps in sheet format, blue TPE, pierceable are suitable for sealing all of our 96 well PCR plates. Ideally suitable for use with individual access and breakable PCR plates.
  • Caps made from TPE mounted on an easily removable backing liner
  • Pierceable with pipette tips
  • Universal fit to 96 well PCR plates
  • Applications: endpoint PCR, storage
  • Not recommended for qPCR
  • Caps perform equivalently to polypropylene cap strips during 25 cycle PCR
  • Can be applied all at once or individually
They are easily pierceable with pipette tips to access samples, and they are easily removable using 1- and 8-way decappers or, alternatively, using Azenta automated plate seal remover if a seal is overlaid on top of the caps.