Handheld Capper/Decapper, Semi-Automated, 8-Channel, Azenta Life Sciences
Supplier: AZENTA US, INC
Certificates
About this item
The semi-automated handheld screw cap decapper, 8-channel provides users with a flexible solution for capping and de-capping 96 format screw caps for low-to-mid throughput environment.
- Lightweight, hand-held, ambidextriouis, semi-automatic single column-based capper and de-capper
- Compatible with 96-SBS rack format screw capped tubes
- Interchangeable cartridges, easy to switch between internally and externally threaded tubes
- Caps a single column in under 4 seconds; caps/de-caps a complete rack of 96 tubes in less than 90 seconds
- All caps are tightened to the same torque to create a secure seal
- Battery operation allows capping/de-capping for more than 40 complete racks
- Docking station for instrument placement/storage and charging also provides contamination-free handling of cap
- Single ‘action’ button operation to de-cap, re-cap and eject caps
- Option to eject caps onto cap carrier using second ‘eject’ button
Includes decapper, docking station, set of power cables, operation manual, cartridges for azenta internal thread and external thread for 96-SBS format racks.
Certifications: CE marked.
Packaging: 1 capper/decapper kit per case.
Frequently Bought Together