AimStrip® Fentanyl Test Strips, Germaine Laboratories
Catalog # 76640-386
Supplier: Germaine Laboratories
Specifications
- Description:AimStrip® Fentanyl Test Strips
- Size:25 Dipsticks/Kit
- Cat. No.:76640-386
- Sensitivity:10 ng/ml
Specifications
About this item
AimStrip® Fentanyl Test Strips are used for the detection of Fentanyl in liquids and powders.
- 25 Strips/Kit
- Sensitivity 10 ng/mL
- Results in 5 minutes
Packaging: Made in USA.
Caution: For Forensic Use Only.