To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:A20 (A-12)
- Clonality:Monoclonal
- Clone:A-12
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Size:6 ml
- Environmentally Preferable:
- Cat. No.:76634-598
Specifications
About this item
A20 Antibody (A-12) is a high quality monoclonal A20 antibody (also designated TNFAIP3 antibody, or TNF a induced protein 3 antibody). A20 is induced by tumor necrosis factor (TNF) and interleukin 1 (IL-1) and acts as a negative regulator of nuclear factor k B (NFkB) gene expression. By inhibiting NFkB activation, A20 plays a critical role in terminating NFkB responses to various stimuli. Although the C-terminal region of A20 contains seven zinc finger domains, only four of these domains are required for in vitro inhibition of TNF-induced NFkB activation. A20 also interacts with several other proteins, such as TRAF2, TRAF6 and IkB kinase (IKK) g protein, and can thereby inhibit cell death. TXBP151, a novel A20-binding protein, may mediate the anti-apoptotic activity of A20. Involved in the negative feedback regulation of signal transduction, A20 and A20-binding proteins may be useful as novel therapeutic tools in the treatment of a variety of diseases.