Anti-Napsin A Mouse Monoclonal Antibody [clone: KCG1.1]
Supplier: Diagnostic Biosystems
Certificates
About this item
Napsin is found in two isoforms, A and B, with highly homologous gene sequences. Napsin A has a molecular weight of 35.0 kDa and is also known as TA02. Napsin A is an aspartic proteinase which is expressed in the lung and involved in processing surfactant protein B (SP-B). It is also expressed in the kidney. This antibody may be a useful tool as a tumor marker for primary lung adenocarcinoma. Napsin expression correlates with the differentiation grade of lung adenocarcinoma.
Specifications
- Catalog No:
- 76633-952
- 76633-956
- 76633-954
- Antigen Name:
- Napsin A
- Napsin A
- Napsin A
- Antibody Type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Clone:
- KCG1.1
- KCG1.1
- KCG1.1
- Reactivity:
- Human
- Human
- Human
- Host:
- Mouse
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- IgG1
Specifications
Frequently Bought Together