Order Entry
United States
ContactUsLinkComponent
Anti-CD23 Mouse Monoclonal Antibody [clone: 1B12]
Anti-CD23 Mouse Monoclonal Antibody [clone: 1B12]
Certificates
Anti-CD23 Mouse Monoclonal Antibody [clone: 1B12]

Specifications

  • Catalog No:
  • 76633-624
  • 76633-622
  • 76633-620
  • Antigen Name:
  • CD23
  • CD23
  • CD23
  • Antibody Type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Clone:
  • 1B12
  • 1B12
  • 1B12
  • Reactivity:
  • Human
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Mouse
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • IgG1 kappa

Specifications

Frequently Bought Together