Order Entry
United States
ContactUsLinkComponent
Beef Extract Powder, Gibco™ (a part of Thermo Fisher Scientific)
Beef Extract Powder, Gibco™ (a part of Thermo Fisher Scientific)
Catalog # 76628-876
CAS Number:  
Beef Extract Powder, Gibco™ (a part of Thermo Fisher Scientific)
Catalog # 76628-876
Supplier Number:  212303
CAS Number:  

Specifications

  • Description:
    Beef extract powder
  • Size:
    500 g
  • Cat. No.:
    76628-876

Specifications

About this item

Beef Extract Powder is a meat extract dried into a powder. This nutritional supplement provides a mixture of peptides, individual amino acids, nucleotides, organic acids, minerals, and vitamins. Additionally, due to differences in the process, beef extract powder has a different molecular weight profile than other beef extract products in our portfolio. 

  • Derived from infusion of beef
  • Provides a rich source of nutrients
  • Free-flowing, homogeneous powder
  • Applications - Intended to replace aqueous infusion of meat in microbiological culture media

Customers have used Beef Extract Powder in a variety of microbial fermentation cell lines including Clostridium sp., Escherichia coli, Bacillus subtillis, Pasteurella multocida, and Streptococcus species to produce numerous animal health vaccines.