ContactUsLinkComponent
Bacto™ Brain Heart Infusion, Gibco™ (a part of Thermo Fisher Scientific)
Bacto™ Brain Heart Infusion, Gibco™ (a part of Thermo Fisher Scientific)
Catalog # 76628-752
CAS Number:  
Bacto™ Brain Heart Infusion, Gibco™ (a part of Thermo Fisher Scientific)
Catalog # 76628-752
Supplier Number:  256110
CAS Number:  
Specifications
  • Description:
    Bacto™ Brain heart infusion, animal origin: Porcine
  • Size:
    10 kg
  • Cat. No.:
    76628-752
Specifications
About this item
Bacto Brain Heart Infusion, porcine, is used for cultivating a wide variety of microorganisms. It was developed as an alternative to the classical Brain Heart Infusion formula and replaces calf brain and beef heart with pork brain and heart.
  • Developed as an alternative to the classical Brain Heart Infusion formula, and replaces calf brains and beef heart with pork brains and heart
  • Formulated with no bovine components to minimize Bovine Spongiform Encephalopathy (BSE) risk
  • Can be used in tests for the verification of fecal streptococci
  • Listed by the National Committee for Clinical Laboratory Standards (NCCLS) as a medium for use in the preparation of microdilution trays for antimicrobial susceptibility testing by the broth microdilution procedure
  • Developed for pharmaceutical and vaccine production
  • Can replace the traditional Brain Heart Infusion depending on organism and production application
Bacto Brain Heart Infusion, porcine, was formulated with no bovine components to minimize bovine spongiform encephalopathy (BSE) risk.