ContactUsLinkComponent
Resurge™ CD Pak, Gibco™ (a part of Thermo Fisher Scientific)
Resurge™ CD Pak, Gibco™ (a part of Thermo Fisher Scientific)
Catalog # 76628-818
CAS Number:  
Resurge™ CD Pak, Gibco™ (a part of Thermo Fisher Scientific)
Catalog # 76628-818
Supplier Number:  670030
CAS Number:  
Specifications
  • Description:
    Resurge™ CD Pak, animal origin-free (chemically defined)
  • Size:
    1 each
  • Cat. No.:
    76628-818
Specifications
About this item
Resurge CD Pak are a diverse set of chemically defined supplement formulations to complement a variety of base media. The CD PAK contains one of each Resurge supplement: CD1, CD2, CD3, CD4, and CD5 at 100 grams each.
  • The Resurge CD Pak contains the following supplements:
  • Resurge CD1 Supplement without glucose and L-glutamine
  • Resurge CD2 Supplement without glucose and L-glutamine
  • Resurge CD3 Supplement without glucose and L-glutamine
  • Resurge CD4 Supplement without glucose and L-glutamine
  • Resurge CD5 Supplement without glucose and L-glutamine
The supplements have been developed using several host cell lines and in various culture systems, including shaker flasks and bench-scale bioreactors. In order to increase versatility of the formulation, glucose and L-glutamine are not included in the final formulations. Therefore, these two components should be added by users and monitored as appropriate.